manufacturer/tradename
Millipore HAHY00010
L × W
10 ft × 1 ft
pore size
0.45 μm
General description
Compatible stains for Immobilon®-NC HAHY membranes are Amido black, India ink, Ponceau-S red, colloidal gold and CPTS. Detection methods like Chromogenic, Radioactive and Fluorescent are suitable.
Nitrocellulose with a small amount of cellulose acetate can be used for improved handling. Water-extractables <5%, including a Triton wetting agent.
Nitrocellulose with a small amount of cellulose acetate can be used for improved handling. Water-extractables <5%, including a Triton wetting agent.
Nitrocellulose with a small amount of cellulose acetate for improved handling. Water-extractables <5%, including a Triton wetting agent. For Southern blots, protein electroblotting, capillary blotting of DNA, RNA, protein.
Application
Immobilon®-NC HAHY membranes is suitable for Southern blotting, Northern blotting, Western blotting and binding assays. It is also suitable for capillary blotting of DNA, RNA and protein.
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Ischemia Reperfusion Iniury in the
Isolated Hemoperfused Bovine
Uterus - a Model for the Investigation
of Anti-Inflammatory Substances?
Isolated Hemoperfused Bovine
Uterus - a Model for the Investigation
of Anti-Inflammatory Substances?
Michael Braun
ALTEX (2003)
Chao Zhang et al.
Cell cycle (Georgetown, Tex.), 14(13), 2100-2108 (2015-05-06)
Osteosarcoma is the most common primary malignant bone tumor and affects a significant portion of pediatric oncology patients. Although surgery and adjuvant chemotherapy confer significant survival benefits, many patients go on to develop metastatic disease, particularly to the lungs, secondary
Hsin-Chieh Lan et al.
Toxicological sciences : an official journal of the Society of Toxicology, 148(1), 26-34 (2015-07-26)
Certain commonly used compounds that interfere with the functions of the endocrine system are classified as endocrine-disrupting chemicals (EDCs). Bisphenol A (BPA) is an EDC that is widely used in food containers. BPA levels in human sera are commonly observed