Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N5881

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide 3′-phosphate sodium salt hydrate

≥90%

Synonym(s):

3′-NADP, 3′-TPN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O17P3 · xNa+ · yH2O
Molecular Weight:
742.40 (anhydrous free acid basis)
UNSPSC Code:
41106305
PubChem Substance ID:

assay

≥90%

impurities

≤5% β-NADP

Storage temp.

−20°C

SMILES string

[Na+].[H]O[H].NC(=O)c1ccc[n+](c1)[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@@H](O)[C@@H]3OP(O)(O)=O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H28N7O17P3.Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-15(31)16(44-46(33,34)35)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)14(30)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;1H2/q;+1;/p-1/t10-,11-,13-,14-,15?,16-,20-,21-;;/m1../s1

Inchi Key

INEQPRUZJJFDHZ-REWCRBICSA-M

Application

β-Nicotinamide adenine dinucleotide 3′-phosphate (3′-NADP) is an analogue of NADP+ used to as a substrate and/or inhibitor to study the specificity and kinetics of NADP-dependent oxidoredutase enzymes.

Other Notes

Analog of β-NADP.

Caution

This is not the common β-NADP required in the usual enzyme systems.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Roy et al.
Journal of biochemical toxicology, 10(2), 111-120 (1995-04-01)
Soybean lipoxygenase-mediated cooxidation of reduced glutathione (GSH) and concomitant superoxide generation was examined. The oxidation of GSH was dependent on the concentration of linoleic acid (LA), GSH, and the enzyme. The optimal conditions to observe maximal enzyme velocity included the
A M Dhalla et al.
Biochemistry, 34(16), 5390-5402 (1995-04-25)
The Escherichia coli MurB gene encoding UDP-N-acetylenolpyruvylglucosamine reductase was expressed to a level of approximately 100 mg/L as a fusion construct with maltose binding protein. Rapid affinity purification, proteolysis, and anion exchange chromatography yielded homogeneous enzyme containing 1 mol/mol bound
Chikako Fukuda et al.
Applied and environmental microbiology, 73(17), 5447-5452 (2007-07-10)
NADP(H) phosphatase has not been identified in eubacteria and eukaryotes. In archaea, MJ0917 of hyperthermophilic Methanococcus jannaschii is a fusion protein comprising NAD kinase and an inositol monophosphatase homologue that exhibits high NADP(H) phosphatase activity (S. Kawai, C. Fukuda, T.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service