Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N5771

Sigma-Aldrich

[O-Methyl-Tyr21]-Neuropeptide Y human

≥97% (HPLC)

Synonym(s):

[Tyr-Me21]-NPY

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C190H287N55O57S
CAS Number:
Molecular Weight:
4285.71
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

Product Name

[O-Methyl-Tyr21]-Neuropeptide Y human, ≥97% (HPLC)

assay

≥97% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NPY(4852)

Amino Acid Sequence

Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr

Biochem/physiol Actions

[O-Methyl-Tyr21]-Neuropeptide Y human is a Neuropeptide Y analog with a high affinity for brain receptors.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Martel et al.
Molecular pharmacology, 38(4), 494-502 (1990-10-01)
A series of fragments and analogues of neuropeptide Y (NPY), both human (hNPY) and porcine (pNPY), were synthesized and tested for their affinities at brain NPY receptor binding sites and their potencies in inhibiting the electrically stimulated twitch response of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service