Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N5640

Sigma-Aldrich

Nα-(3-Nitro-2-pyridinesulfenyl)-N-im-tosyl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17N5O6S2
CAS Number:
Molecular Weight:
463.49
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)n2cnc(CC(NSc3ncccc3N(=O)=O)C(O)=O)c2

InChI

1S/C18H17N5O6S2/c1-12-4-6-14(7-5-12)31(28,29)22-10-13(20-11-22)9-15(18(24)25)21-30-17-16(23(26)27)3-2-8-19-17/h2-8,10-11,15,21H,9H2,1H3,(H,24,25)

InChI key

HREPATZNEVVIAA-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service