Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N5384

Sigma-Aldrich

Nervonoyl coenzyme A

≥85%

Synonym(s):

cis-15-Tetracosenoyl Coenzyme A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H80N7O17P3S
CAS Number:
Molecular Weight:
1116.14
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.26

Quality Level

assay

≥85%

storage temp.

−20°C

SMILES string

[P](=O)(O[P](=O)(OCC(C(O)C(=O)NCCC(=O)NCCSC(=O)CCCCCCCCCCCCC\C=C\CCCCCCCC)(C)C)O)(OCC1OC(C(C1O[P](=O)(O)O)O)[n]2c3ncnc(c3nc2)N)O

InChI

1S/C45H80N7O17P3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-36(54)73-29-28-47-35(53)26-27-48-43(57)40(56)45(2,3)31-66-72(63,64)69-71(61,62)65-30-34-39(68-70(58,59)60)38(55)44(67-34)52-33-51-37-41(46)49-32-50-42(37)52/h11-12,32-34,38-40,44,55-56H,4-10,13-31H2,1-3H3,(H,47,53)(H,48,57)(H,61,62)(H,63,64)(H2,46,49,50)(H2,58,59,60)/b12-11+

InChI key

QHZAQVTVYPHLKK-VAWYXSNFSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service