Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N5287

Sigma-Aldrich

Monoclonal Anti-Nuf2 antibody produced in mouse

~2 mg/mL, clone 28-37, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

28-37, monoclonal

form

buffered aqueous solution

mol wt

antigen ~52 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using total cell extract of A549 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUF2(83540)

General description

Monoclonal Anti-Nuf2 (mouse IgG1isotype) is derived from the hybridoma 28-37 produced by the fusion of mouse myeloma cells (PAI cells) and splenocytes from BALB/c mice. NUF2 gene is mapped to human chromosome 1q23.3. NUF2 protein is a component of the kinetochore complex.
NDC80 kinetochore complex component, NUF2 is also called as cell division associated 1 (CDCA1).

Specificity

Monoclonal Anti-Nuf2 recognizes human Nuf2.

Immunogen

recombinant full length human Nuf2.

Application

Monoclonal Anti-Nuf2 antibody produced in mouse may be used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry
  • immunoprecipitation

Biochem/physiol Actions

Downregulation of the NUF2 protein by RNA interference results in the failure of the kinetochores to form attachments to the spindle microtubules. As a consequence, cells are blocked in the prometaphase stage with an active spindle checkpoint and undergo cell death.
NDC80 kinetochore complex component, NUF2 is highly expressed in glioma. It has important roles in attachment of the microtubule to the kinetochore and in chromosome segregation. NUF2 is also involved in enhancing cell growth in hepatocellular carcinoma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in “frost-free” freezers is also not recom-mended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meirong Zang et al.
Oncotarget, 6(33), 34276-34287 (2015-09-30)
Multiple myeloma (MM) is a genetically heterogeneous disease with diverse clinical characteristics and outcomes. Recently, multiplex ligation-dependent probe amplification (MLPA) has emerged as an effective and robust method for the detection of cytogenetic aberrations in MM patients. In the present
Qiang Liu et al.
Asian Pacific journal of cancer prevention : APJCP, 15(20), 8623-8629 (2014-11-07)
As an important component of the NDC80 kinetochore complex, NUF2 is essential for kinetochore-microtubule attachment and chromosome segregation. Previous studies also suggested its involvement in development of various kinds of human cancers, however, its expression and functions in human hepatocellular
S-K Huang et al.
Cellular and molecular biology (Noisy-le-Grand, France), 60(4), 30-36 (2014-12-08)
Glioma is one of the most commonly malignant brain tumors. Current therapies for glioma have failed to achieve satisfactory results, which necessitates the development of novel molecular therapies. In the current study, we aimed to investigate the role of NUF2
Peng Hu et al.
Bioscience reports, 35(1), doi:10-doi:10 (2014-11-06)
NUF2 (NUF2, Ndc80 kinetochore complex component) plays an important role in kinetochore-microtubule attachment. It has been reported that NUF2 is associated with multiple human cancers. However, the functional role of NUF2 in pancreatic cancer remains unclear. In this study, we
Jennifer G DeLuca et al.
The Journal of cell biology, 159(4), 549-555 (2002-11-20)
Identification of proteins that couple kinetochores to spindle microtubules is critical for understanding how accurate chromosome segregation is achieved in mitosis. Here we show that the protein hNuf2 specifically functions at kinetochores for stable microtubule attachment in HeLa cells. When

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service