N4415
NGA2F Glycan
from Porcine
Synonym(s):
(GlcNAc)2Man3(Fuc)(GlcNAc)2, Asialo, agalacto, biantennary N-glycan, core fucosylated, FA2 N-glycan, Mannotriose-(fucosyl-di-[N-acetylglucosamine]), bis(N-acetylglucosaminyl)-
Select a Size
Select a Size
About This Item
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service