Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N4285

Sigma-Aldrich

Anti-NCS-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Neuronal Calcium Sensor-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~20 kDa

species reactivity

rat

technique(s)

western blot: 1 μg/mL using rat brain extract

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FREQ(23413)
mouse ... Freq(14299)
rat ... Freq(65153)

General description

Neuronal calcium sensor 1 (NCS-1) is a Ca2+ signaling protein.

Immunogen

synthetic peptide correspopnding to amino acid residues 161-190 of rat NCS-1. This sequence is completely conserved in mouse, rat, chicken, and human NCS-1. There is a single amino acid substitution in this region for Xenopus NCS-1.

Biochem/physiol Actions

Neuronal calcium sensor 1 (NCS-1) responds to changes in Ca2+ levels and has a crucial function in memory and learning. NCS-1 is involved in many regulatory pathways in neurons, and associates with kinases and G-protein coupled receptors.

Physical form

Solution in phosphate buffered saline containing 1 mg/ml bovine serum albumin and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luciana Estefani Drumond et al.
Brain research bulletin, 88(4), 385-391 (2012-04-24)
Physical activity has been proposed as a behavioral intervention that improves learning and memory; nevertheless, the mechanisms underlying these health benefits are still not well understood. Neuronal Calcium Sensor-1 (NCS-1) is a member of a superfamily of proteins that respond
Sravan Pandalaneni et al.
The Journal of biological chemistry, 290(30), 18744-18756 (2015-05-17)
Neuronal calcium sensor-1 (NCS-1) is the primordial member of the neuronal calcium sensor family of EF-hand Ca(2+)-binding proteins. It interacts with both the G-protein-coupled receptor (GPCR) dopamine D2 receptor (D2R), regulating its internalization and surface expression, and the cognate kinases
Ping K Yip et al.
PLoS biology, 8(6), e1000399-e1000399 (2010-06-30)
Following trauma of the adult brain or spinal cord the injured axons of central neurons fail to regenerate or if intact display only limited anatomical plasticity through sprouting. Adult cortical neurons forming the corticospinal tract (CST) normally have low levels

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service