Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N4165

Sigma-Aldrich

Neuroendocrine Regulatory Peptide-1 (human)

Synonym(s):

NERP-1 (Human)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

form

lyophilized powder

Quality Level

color

white

solubility

water, double-distilled: soluble

storage temp.

−20°C

Amino Acid Sequence

H-Arg-Pro-Glu-Ser-Ala-Leu-Leu-Gly-Gly-Ser-Glu-Ala-Gly-Glu-Arg-Leu-Leu-Gln-Gln-Gly-Leu-Ala-Gln-Val-Glu-Ala-NH2

General description

NERP1/2 peptides are responsible to a variety of physiological processes:
a. Modulation of body fluid homeostasis.
b. Suppression of basal and angiotensin II-induced vasopressin release from hypothalamic organ.
c. They were shown to evoke calcium responses in the pituitary gland and hypothalamus.

Physical form

Lyophilized from 0.1% aqueous TFA

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Matsuo et al.
Regulatory peptides, 163(1-3), 43-48 (2010-05-18)
Neuroendocrine regulatory peptides NERP-1 and NERP-2 are novel amidated peptides derived from VGF, a polypeptide secreted from neurons and endocrine cells through a regulated pathway. To explore the localization of NERPs in human tissues, we performed immunohistochemistry analysis on tissues
K Toshinai et al.
Cellular and molecular life sciences : CMLS, 66(11-12), 1939-1945 (2009-02-06)
Neuroendocrine regulatory peptides (NERP)-1 and NERP-2 are derived from distinct regions of VGF, a neurosecretory protein that was originally identified as a product of a nerve growth factor-responsive gene in rat PC12 cells. The amino acid length of human NERP-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service