Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

N3631

Sigma-Aldrich

2-Naphthoxyacetic acid sodium salt

crystalline

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$143.45

$143.45

List Price$151.00Save 5%
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$143.45

About This Item

Linear Formula:
C12H9O3Na
CAS Number:
Molecular Weight:
224.19
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

$143.45

List Price$151.00Save 5%
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

form

crystalline

Quality Level

color

white

application(s)

agriculture

SMILES string

[Na].OC(=O)COc1ccc2ccccc2c1

InChI

1S/C12H10O3.Na.H/c13-12(14)8-15-11-6-5-9-3-1-2-4-10(9)7-11;;/h1-7H,8H2,(H,13,14);;

InChI key

HYGDEOFLJJUVKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service