Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N3414

Sigma-Aldrich

NSC348884 hydrate

Synonym(s):

N1,N2-bis((3-imino-6-methyl-3H-indol-2-yl)methyl)-N1,N2-bis((6-methyl-1H-benzo[d]imidazol-2-yl)methyl)ethane-1,2-diamine hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$138.00
25 MG
$388.00

$138.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$138.00
25 MG
$388.00

About This Item

Empirical Formula (Hill Notation):
C38H40N10 · xH2O
CAS Number:
Molecular Weight:
636.79 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$138.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

form

solid

Quality Level

color

white

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O.Cc1ccc2[nH]c(CN(CCN(Cc3nc4cc(C)ccc4[nH]3)Cc5nc6cc(C)ccc6[nH]5)Cc7nc8cc(C)ccc8[nH]7)nc2c1

InChI

1S/C38H40N10.H2O/c1-23-5-9-27-31(15-23)43-35(39-27)19-47(20-36-40-28-10-6-24(2)16-32(28)44-36)13-14-48(21-37-41-29-11-7-25(3)17-33(29)45-37)22-38-42-30-12-8-26(4)18-34(30)46-38;/h5-12,15-18H,13-14,19-22H2,1-4H3,(H,39,43)(H,40,44)(H,41,45)(H,42,46);1H2

InChI key

AFSOHVUGOYTFCG-UHFFFAOYSA-N

Biochem/physiol Actions

NSC 348884 is a putative inhibitor of nucleophosmin (NPM).
NSC 348884 is a putative inhibitor of nucleophosmin (NPM). NSC 348884 inhibits NMP oligomer formation, up-regulates p53 and induces apoptosis.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachy Abraham et al.
Journal of proteome research, 16(11), 4144-4155 (2017-09-30)
Chikungunya virus (CHIKV), a positive-stranded RNA virus, can cause neurological complications by infecting the major parenchymal cells of the brain such as neurons and astrocytes. A proteomic analysis of CHIKV-infected human astrocytic cell line U-87 MG revealed tight functional associations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service