Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N3287

Sigma-Aldrich

Monoclonal Anti-Neuropilin-1 antibody produced in mouse

clone 130603, purified immunoglobulin, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

130603, monoclonal

form

lyophilized powder

species reactivity

rat

technique(s)

indirect ELISA: 0.5-1 μg/mL using rat Neuropilin-1

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

During development, neuropilin-1 (NRP-1) is expressed in arteries. It is also a co-receptor of semaphorin-3a.

Specificity

Shows no cross-reactivity with recombinant rat neuropilin-2.

Immunogen

purified, NS0-derived, recombinant rat neuropilin-1.

Biochem/physiol Actions

Neuropilin-1 (NRP-1) has been shown to play an important role in vascular and neural functions. Neuropilin 1 (Nrp1) acts as a coreceptor for VEGF receptor 2 (VEGFR2) and increases the binding of VEGF165 to VEGFR2- and VEGF165-mediated chemotaxis. In transgenic mice, Nrp1 is implicated in embryonic vascular development. Elevated expression of the gene leads to the formation of excess capillaries and haemorrhaging.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Role of Neuropilin in Vascular and Tumor Biology
Klagsbrun M
Advances in Experimental Medicine and Biology, 515, 33-48 (2002)
Michiko Hayashi et al.
Neuroscience research, 69(1), 17-24 (2010-10-05)
Semaphorins, one of the repulsive axonal guidance factors during development, are produced under pathological conditions in adult animals. In the neuropathic pain state associated with peripheral nerve injury, synaptic reorganization occurs in spinal cord dorsal horn. In the present study
Katarzyna A Uniewicz et al.
The Journal of biological chemistry, 286(1), 12-23 (2010-10-20)
Neuropilin-1 (NRP-1) is present on the cell surface of endothelial cells, or as a soluble truncated variant. Membrane NRP-1 is proposed to enhance angiogenesis by promoting the formation of a signaling complex between vascular endothelial growth factor-A(165) (VEGF-A(165)), VEGF receptor-2
Ling Fu et al.
World journal of gastroenterology, 18(36), 5034-5041 (2012-10-11)
To elucidate the role of neuropilin-1 (Nrp-1) and semaphorin 3A (Sema3A) in sinusoidal remodeling during liver regeneration in rats. Male Wistar/ST rats at 7 wk of age, weighing about 200 g, were used for all animal experiments. In vivo, at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service