Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N2661

Sigma-Aldrich

Monoclonal Anti-NAK (NFκB-Activating Kinase) antibody produced in mouse

~2.5 mg/mL, clone NAK369, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NAK369, monoclonal

form

buffered aqueous solution

mol wt

antigen 84 kDa

species reactivity

human, rat, chicken

concentration

~2.5 mg/mL

technique(s)

indirect ELISA: suitable
microarray: suitable
western blot: 1-2 μg/mL using 293T cells transfected with a human NAK expression vector

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBK1(29110)
rat ... Tbk1(299827)

General description

Nuclear factor κB (NF-κB) is a transcription factor that regulates a wide range of functions such as cell growth, angiogenesis, immunological responses and cell death. This protein has also been implicated in inflammation and carcinogenesis. Overexpression of NF-κB has been associated with breast and esophageal cancer . Monoclonal Anti-NAK antibody is specific for human, rat and chicken NAK.

Immunogen

human NAK fused to GST.

Application

Monoclonal Anti-NAK (NFκB-Activating Kinase) antibody can be used for western blot (1:250) and immunohistochemistry (1:50) assays in human mucosal tissues . The antibody is also suitable for use in indirect ELISA and microarrays.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chae Hyeong Lee et al.
BioFactors (Oxford, England), 29(1), 19-35 (2007-07-06)
Nuclear factor kappaB (NF-kappaB), a transcription factor, plays an important role in carcinogenesis as well as in the regulation of immune and inflammatory responses. NF-kappaB induces the expression of diverse target genes that promote cell proliferation, regulate apoptosis, facilitate angiogenesis
James T Wu et al.
The Journal of surgical research, 123(1), 158-169 (2005-01-18)
The nuclear factor kappaB (NFkappaB) superfamily of eukaryotic transcription factors plays an important role in carcinogenesis. NF-kappaB and its regulators are linked to various signal transduction pathways as well as transcriptional activation events that mediate critical stages of cell proliferation.
Norihisa Uemura et al.
Cancer science, 100(9), 1612-1622 (2009-06-30)
To identify the molecular background of esophageal cancer, we conducted a proteomics study using an antibody microarray consisting of 725 antibodies and surgical specimens from three cases. The microarray analysis identified 24 proteins with aberrant expression in esophageal cancer compared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service