Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N2290

Sigma-Aldrich

NP603

≥98% (HPLC)

Synonym(s):

(Z)-3-(5-(6-(3,5-Dimethoxyphenyl)-2-oxo-1,2-dihydro-indol-3-ylidenemethyl)-2,4-dimethyl-1H-pyrrol-3-yl)-propionic acid, 5-[[6-(3,5-Dimethoxyphenyl)-1,2-dihydro-2-oxo-3H-indol-3-ylidene]methyl]-2,4-dimethyl-1H-pyrrole-3-propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N2O5
CAS Number:
Molecular Weight:
446.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

orange-red

solubility

DMSO: ≥30 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc(OC)cc(c1)-c2ccc3c(NC(=O)\C3=C/c4[nH]c(C)c(CCC(O)=O)c4C)c2

InChI

1S/C26H26N2O5/c1-14-20(7-8-25(29)30)15(2)27-23(14)13-22-21-6-5-16(11-24(21)28-26(22)31)17-9-18(32-3)12-19(10-17)33-4/h5-6,9-13,27H,7-8H2,1-4H3,(H,28,31)(H,29,30)/b22-13-

InChI key

ZIZSHSPAAJIYOD-XKZIYDEJSA-N

Biochem/physiol Actions

NP603 is a cell-permeable inhibitor of PDGFRβ, FGFR1 and VEGFR2 that binds to ATP pocket.
NP603 is a cell-permeable inhibitor of PDGFRβ, FGFR1 and VEGFR2 that binds to ATP pocket. NP603 is a very weak inhibitor of EGFR (IC50 >100 μM).

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the FGFR and PDGFR pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to FGFR for your research needs.

We offers many products related to PDGFR for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service