Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N2034

Sigma-Aldrich

CR 2249

Synonym(s):

(S)-4-Amino-5-[(4,4-dimethylcyclohexyl)amino]-5-oxopentanoic acid, Nebostinel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24N2O3
CAS Number:
Molecular Weight:
256.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CC1(C)CCC(CC1)NC(=O)[C@@H](N)CCC(O)=O

InChI

1S/C13H24N2O3/c1-13(2)7-5-9(6-8-13)15-12(18)10(14)3-4-11(16)17/h9-10H,3-8,14H2,1-2H3,(H,15,18)(H,16,17)/t10-/m0/s1

InChI key

VCRGLZYPNNAVRP-JTQLQIEISA-N

Biochem/physiol Actions

Facilitates glycine action at NMDA glutamate receptors; may identify a separate allosteric modulatory site on the NMDA receptor; putative cognition enhancer.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pittaluga et al.
The Journal of pharmacology and experimental therapeutics, 290(1), 423-428 (1999-06-25)
Some cognition enhancers were previously shown to potently prevent antagonism of the N-methyl-D-aspartate (NMDA)-evoked release of norepinephrine (NE) brought about in slices of rat hippocampus by kynurenic acid, an endogenous NMDA receptor blocker. We have examined the impact of putative
M Lanza et al.
Neuropharmacology, 36(8), 1057-1064 (1997-08-01)
The effects of (S)-4-amino-5-[(4,4-dimethylcyclohexyl)amino]-5-oxo-pentanoic acid ((S)CR 2249), a new chemical entity selected among a series of glutamic acid derivatives, were investigated on N-methyl-D-aspartate (NMDA)-evoked release of [3H]noradrenaline from rat hippocampal slices. (S)CR 2249 facilitated glycine-mediated reversion of kynurenate antagonism at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service