Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N190

Sigma-Aldrich

[D-Tyr27,36, D-Thr32]-Neuropeptide Y fragment 27-36, rat

>97%, lyophilized powder

Synonym(s):

[D-Tyr27,36, D-Thr32]-NPY Fragment 27-36, rat, D-Tyr-Ile-Asn-Leu-Ile-D-Thr-Arg-Gln-Arg-D-Tyr-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C61H98N19O15
Molecular Weight:
1337.55
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

>97%

form

lyophilized powder

color

white

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](N)Cc1ccc(O)cc1)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@H]([C@H](C)O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@H](Cc2ccc(O)cc2)C(N)=O

InChI

1S/C61H99N19O15/c1-8-31(5)47(78-51(87)38(62)27-34-14-18-36(82)19-15-34)57(93)77-44(29-46(64)85)55(91)76-43(26-30(3)4)56(92)79-48(32(6)9-2)58(94)80-49(33(7)81)59(95)74-40(13-11-25-71-61(68)69)52(88)73-41(22-23-45(63)84)54(90)72-39(12-10-24-70-60(66)67)53(89)75-42(50(65)86)28-35-16-20-37(83)21-17-35/h14-21,30-33,38-44,47-49,81-83H,8-13,22-29,62H2,1-7H3,(H2,63,84)(H2,64,85)(H2,65,86)(H,72,90)(H,73,88)(H,74,95)(H,75,89)(H,76,91)(H,77,93)(H,78,87)(H,79,92)(H,80,94)(H4,66,67,70)(H4,68,69,71)/t31-,32-,33-,38+,39-,40-,41-,42+,43-,44-,47-,48-,49+/m0/s1

InChI key

ZSPXOUGXONWRQC-FHDPKJCZSA-N

Biochem/physiol Actions

[D-Tyr27,36, D-Thr32]-Neuropeptide Y fragment 27-36, rat is neurotransmitter.

Caution

Hygroscopic

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M VanNess et al.
Brain research, 821(2), 263-269 (1999-03-05)
We hypothesized that hypothalamic NPYergic mechanisms mediate the blood pressure lowering effect of caloric restriction in hypertensive rats. Aortic coarctation-induced (AC) hypertensive rats (n=25) were assigned to either an ad libitum fed control group (AL) or food restricted group (FR;
R D Myers et al.
Brain research bulletin, 37(3), 237-245 (1995-01-01)
Neuropeptide Y (NPY) produces a vigorous feeding response in several species when it is injected into hypothalamic structures involved in eating behavior. The purpose of this study was to determine whether a unique carboxy terminal fragment of NPY would alter
Neuronal and non-neuronal modulation of sympathetic neurovascular transmission
Macarthur, H.
Acta Physiol., 203, 37-45 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service