N1780
Neurotrophin-4 human
recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Synonym(s):
NTF4, NT-4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
recombinant
expressed in baculovirus infected Sf21 cells
assay
>97% (SDS-PAGE)
form
lyophilized powder
potency
0.3-15.0 ng/mL EC50
mol wt
14 kDa
packaging
pkg of 5 μg
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
UniProt accession no.
Storage temp.
−20°C
Gene Information
human ... NTF4(4909)
Biochem/physiol Actions
Neurotrophin-4 (also called NT-4/5 or NT-5) is a 14 kDa member of the neurotrophin family. NT-4 shares 95% sequence homology between human and rat. NT-4 from human, rat, and Xenopus are active on chick dorsal root ganglia. NT-4 is expressed in many tissues and major brain regions. NT-4 promotes survival and differentiation of various cells in culture including spinal neuons, basal forebrain cholinergic neurons, hippocampal neurons, cerebellar granule cells, embryonic dopaminergic neruons of mesencephalon, noradrenergic neurons of the locus coeruleus, dopaminergic, GABAergic, and serotoninergic neurons of the substantia nigra, and embryonic trigeminal and jugular neurons.
Physical form
Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% TFA, containing 50 μg bovine serum albumin per 1 μg of cytokine.
Analysis Note
The biological activity is measured in a cell proliferation assay using a TrkB-transfected cell line BaF-TrkB-BD.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T Timmusk et al.
The European journal of neuroscience, 5(6), 605-613 (1993-06-01)
The neurotrophin gene family includes four structurally related proteins with neurotrophic activities. Two of them, nerve growth factor and brain-derived neurotrophic factor (BDNF), have been studied in detail and information has recently emerged on the expression and function of the
Daniel Todd et al.
PloS one, 9(2), e87923-e87923 (2014-02-08)
Huntington's disease (HD) is a devastating, genetic neurodegenerative disease caused by a tri-nucleotide expansion in exon 1 of the huntingtin gene. HD is clinically characterized by chorea, emotional and psychiatric disturbances and cognitive deficits with later symptoms including rigidity and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service