Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1411

Sigma-Aldrich

Ninhydrin Fixer Spray Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

storage temp.

2-8°C

Application

Fixative for ninhydrin chromatograms. Product does not include spray bottle.

Other Notes

Acidified ethanol containing 1% (v/v) saturated cupric nitrate.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Ox. Liq. 1 - Skin Irrit. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

55.0 °F - closed cup

flash_point_c

12.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service