N0790
NDSB 221
≥97% (TLC)
Synonym(s):
3-(1-Methylpiperidinio)-1-propanesulfonate, 3-(1-Methylpiperidinium)-1-propane sulfonate
About This Item
Recommended Products
description
zwitterionic
assay
≥97% (TLC)
form
solid
mol wt
221.32 g/mol
color
white
solubility
H2O: 50 mg/mL, clear, colorless
General description
Application
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service