Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N0535

Sigma-Aldrich

N-(NBD-Aminocaproyl)-L-α-phosphatidyl-DL-glycerol, α-myristoyl sodium salt

≥95%, solid

Synonym(s):

1-Tetradecanoyl-2-[6-[(7-nitro-2-,1,3-benzoxadiazol-4-yl)amino]hexanoyl]-sn-glycerol-3-phospho-rac-(1-glycerol) sodium salt, 14:0-6:0 NBD PG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H52N4NaO13P
Molecular Weight:
754.74
MDL number:
UNSPSC Code:
12352211

assay

≥95%

form

solid

shipped in

dry ice

storage temp.

−20°C

General description

Fluorescently labeled fatty acid phospholipid membrane probe.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service