Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N0379

Sigma-Aldrich

Nucleoside 5′-Diphosphate Kinase from baker′s yeast (S. cerevisiae)

lyophilized powder

Synonym(s):

ATP:Nucleoside diphosphate phosphotransferase, NDPK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

1000-2000 units/mg protein

composition

Protein, 10-40% modified Warburg-Christian

UniProt accession no.

storage temp.

−20°C

Gene Information

bakers yeast ... YNK1(853798)

General description

Nucleoside 5′-diphosphate kinase is a cytosolic enzyme. [1]

Application

Nucleoside 5′-diphosphate kinase has been used in a study to examine a possible intracellular activity of the drug disodium cromoglycate in mast cells. [1] It has also been used in a study to investigate protein synthesis in rabbit reticulocytes. [2]

Biochem/physiol Actions

Nucleoside 5′-diphosphate kinase from Saccharomyces cerevisiae is found highly expressed in the cytoplasm. It affects DNA synthesis, in part, by binding to Cdc8p. [3]
Transfers one phosphate from many nucleoside triphosphates to many nucleoside diphosphates. No activity when UMP is substituted for the diphosphate.

Unit Definition

One unit will convert 1.0 μmole each of TDP and ATP to TTP and ADP per min at pH 7.6 at 25 °C in a coupled system with PK/LDH.

Physical form

Lyophilized, essentially sulfate-free powder. Contains sodium citrate with a trace of magnesium and EDTA salts.

Reconstitution

Reconstitute with deionized water. Solution is believed to be stable for over 1 year while frozen.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temporal and spatial distributions of yeast nucleoside diphosphate kinase activities and its association with the Cdc8p.
Zhang, SQ.
Cellular and Molecular Biology, 41, 333-346 (1995)
Takumi Chinen et al.
Nature communications, 6, 8722-8722 (2015-10-28)
Inhibitors of microtubule (MT) assembly or dynamics that target α/β-tubulin are widely exploited in cancer therapy and biological research. However, specific inhibitors of the MT nucleator γ-tubulin that would allow testing temporal functions of γ-tubulin during the cell cycle are
R Roy et al.
Biochemical and biophysical research communications, 122(3), 1418-1425 (1984-08-16)
Under standard conditions, in the presence of GTP, highly purified eIF-2 and Co-eIF-2 factor preparations efficiently stimulated AUG-codon dependent but not physiological mRNA-dependent Met-tRNAf binding to 40S ribosomes. Replacement of GTP by a nonhydrolyzable GTP analog, GMP-PNP, in the above
S Hemmerich et al.
Biochemistry, 30(6), 1523-1532 (1991-02-12)
Type I Fc epsilon receptor (Fc epsilon RI) mediated Ca2+ uptake and secretion of rat serosal mast cells have been shown to be inhibited by disodium 1,3-bis [(2'-carboxylatochromon-5'-yl) oxy]-2-hydroxypropane (disodium cromoglycate, DSCG), which is widely employed in the treatment of
J A Stahl et al.
Cancer research, 51(1), 445-449 (1991-01-01)
Reduced RNA and/or protein levels corresponding to the murine nm23-1 and human nm23-H1 complementary DNA clones have been correlated with high tumor metastatic potential in several rodent model systems and human breast carcinomas. We report the identification of a second

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service