MTOXHRSUP
HepaRG™ Recovery Medium Supplement
sterile-filtered
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Sigma has developed a variety of specialized media which are intended to provide optimal performance when utilized with specific cell lines. This product consists of 62.5 mL of HepaRG™ Recovery Medium supplement which is added to basal media and other components listed below to produce 500 mL of complete media needed for recovery and Sandwich culture of HepaRG™ cells. The final HepaRG™ recovery medium was developed specifically for use with Sigma′s human HepaRG™ control 5F Clone cells and various knockouts (KO).
Preparation Note
Prepare Recovery Medium by supplementing 500 mL of William′s E Medium (W1878) ) with 5 mL of Penicillin-Streptomycin solution (P4333), 5 mL of GlutaMAX (Life Technologies 35050-061), and the entire Recovery Medium Supplement (MTOXHRSUP). Complete Medium can be stored at 2–8 °C for up to 1 month. This medium is formulated for use with a 5% CO2 in air atmosphere.
Other Notes
Upon receiving a shipment of supplement(s) it is important the end user gives the shipment attention without delay. To ensure the highest quality of the supplement(s), upon receipt supplements should be immediately stored at –20 °C. Upon thaw, medium supplement will be clear to turbid. Upon addition to the Williams′ E Medium, medium should be clear.
Legal Information
6538 is a trademark of American Type Culture Collection
HepaRG is a trademark of BioPredic International company
signalword
Warning
hcodes
Hazard Classifications
Repr. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service