Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MTOXHPSUP

Sigma-Aldrich

HepaRG Pre-Induction Medium Supplement

sterile-filtered

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Sigma has developed a variety of specialized media which are intended to provide optimal performance when utilized with specific cell lines. This product consists of 59 mL of HepaRG Pre-Induction Medium supplement which is added to basal media and other components listed below to produce 500 mL of complete media needed for induction assays with HepaRG cells. The final HepaRG pre-induction medium was developed specifically for use with Sigma′s human HepaRG control 5F Clone cells and various knockouts (KO).

Preparation Note

Prepare Pre-Induction Medium by supplementing 500 mL of William′s E Medium (W1878) ) with 5 mL of Penicillin-Streptomycin solution (P4333), 5 mL of GlutaMAX (Life Technologies 35050-061), and the entire Pre-Induction Medium Supplement (MTOXHPSUP). Complete Medium can be stored at 2–8 °C for up to 1 month. This medium is formulated for use with a 5% CO2 in air atmosphere.

Other Notes

Upon receiving a shipment of supplement(s) it is important the end user gives the shipment attention without delay. To ensure the highest quality of the supplement(s), upon receipt supplements should be immediately stored at –20 °C. Upon thaw, medium supplement will be clear to turbid. Upon addition to the Williams′ E Medium, medium should be clear.

Legal Information

HepaRG is a trademark of BioPredic International company

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service