MSST0009
SILu™Prot MAPK1 Mitogen Activated Protein Kinase 1 human
recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N -labeled
Synonym(s):
Extracellular signal-regulated kinase 2 (Erk2), Mitogen-activated protein kinase 1 (MAPK1)
About This Item
Recommended Products
General description
Biochem/physiol Actions
Sequence
Physical form
Legal Information
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service