Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MI00300

Sigma-Aldrich

MISSION® microRNA Mimic

Human miRNA Mimic Library, miRBase version 21 library

Synonym(s):

miRNA mimic library

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

MISSION®

form

solid

storage temp.

−20°C

General description

Human miRNA mimic library, based on miRBase Version 21 from the Sanger Institute, supplied as 2,754 miRNA mimics at 0.25nmol each in 96-well plate format.
The ready-to-use MISSION miRNA mimics are small, double-stranded RNA molecules designed to mimic endogenous mature miRNA molecules when introduced into cells. miRNA are known to regulate gene expression in a variety of manners, including translational repression, mRNA cleavage and deadenylation. MISSION miRNA Mimics, a member of MISSION RNAi product family, provides miRNA researchers with a range of options from individual MISSION mimics to a full library of human miRNA mimics based on latest version of miRBase (currently hosted by the University of Manchester, previously hosted by the Sanger Institute).

Features and Benefits

  • Unique MISSION miRNA mimic design significantly reduces possible sense strand off target effects.
  • Novel MISSION miRNA mimic design has been functionally tested for knockdown efficiency against natural miRNA targets.
  • Library contains all human mature miRNAs in miRBase version 21.
  • Coverage includes 2,754 miRNA mimics
  • Optimized and ready for transfection.
  • Library is in 96-well format, and 384-well plate format is available upon request.
  • Negative controls included in each plate of the miRNA mimic library. 
  • Available as a whole human library and individual miRNA targets.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service