Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MED-HHTM

Sigma-Aldrich

Human Hepatocyte Thawing Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

shipped in

dry ice

Storage temp.

−20°C

General description

Human Hepatocyte Thawing Medium has been developed for thawing and clarifying cryovials of frozen hepatocytes. Upon receiving a shipment of supplement(s) it is important the end user gives the shipment attention without delay. To ensure the highest quality of the supplement(s), upon receipt supplements should be immediately stored at –20 °C.

Application

See technical bulletin for protocols for use with LifeNet Health® Primary Human Hepatocytes.

Legal Information

LifeNet Health is a registered trademark of LifeNet Health

configured for

Product No.
Description
Pricing

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service