Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MED-HHCMSP

Sigma-Aldrich

Human Hepatocyte Culture Medium Supplement Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

shipped in

dry ice

storage temp.

−20°C

General description

The supplement pack is a 3 component kit for use with human hepatocyte culture medium to prepare required volumes of complete media. Upon receiving a shipment of supplement(s) it is important the end user gives the shipment attention without delay. To ensure the highest quality of the supplement(s), upon receipt supplements should be immediately stored at –20 °C.

Application

For use with Human Hepatocyte Culture Medium. See technical bulletin for LifeNet Health® primary human hepatocytes for protocols.

Legal Information

LifeNet Health is a registered trademark of LifeNet Health

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Repr. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service