Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MAK351

Sigma-Aldrich

Diamine Oxidase Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

The new Diamine Oxidase Assay Kit, MAK526, is now available! Diamine Oxidase (DAO) is an important regulator of polyamine levels in human physiology. DAO metabolizes the oxidative conversion of biogenic polyamines such as putrescine and cadaverine into aldehydes. Biogenic amines are known regulators of important biological pathways such as cell growth and signal transduction, placing DAO in an important regulatory position in maintaining cell viability. Putrescine itself is also an activator of c-jun and c-fos expression, further linking this diamine with potential roles in cancer development. DAO is notably involved in regulating growth and differentiation of intestinal mucosa, bone marrow, and other rapidly proliferating tissue. In addition to its role in tissue growth, DAO activity is important in metabolism. Loss of diamine oxidase activity can lead to histamine intolerance and histamine poisoning.

Suitability

Suitable for the measurement of DAO activity in various tissues and cell extracts as well as the characterization of DAO activity of purified recombinant enzyme.

Principle

The Diamine Oxidase Activity Assay Kit provides a straightforward method to determine DAO activity of tissue lysates as well as recombinant enzymes, with a detection limit of less than 1 pmole/minute of activity. In the assay, DAO converts the provided substrate, yielding an intermediate and H2O2. H2O2 is then utilized by the DAO Enzyme Mix to generate fluorescence (Ex/Em = 535/587 nm) from the DAO Probe.

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

188.6 °F

flash_point_c

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service