Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK295

Sigma-Aldrich

Monoamine Oxidase-A Inhibitor Screening Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

detection method

fluorometric

relevant disease(s)

neurological disorders

Storage temp.

−20°C

General description

The new Monoamine Oxidase (MAO) Inhibitor Screening Kit, MAK520, is now available! Monoamine oxidases (MAO, EC 1.4.3.4) are a family of enzymes that can oxidize a wide variety of endogenous primary amines. Two isoforms, MAO-A and MAO-B, have been identified based on their substrate, inhibitor specificity and tissue localization. MAO-B can oxidize primary amines, but its list of specific substrates (i.e. benzylamine, phenylethylamine) is more limited compared to MAO-A.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for screening/studying/characterizing MAO-A inhibitors.

Principle

This MAO-A Inhibitor Screening Kit offers a rapid, simple, sensitive, and reliable test suitable for high-throughput screening of MAO-A inhibitors. The assay is based on the fluorometric detection of hydrogen peroxide (H2O2), one of the by-products generated during the oxidative deamination of the MAO substrate (tyramine).

Pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service