Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK289

Sigma-Aldrich

Purine Nucleoside Phosphorylase Activity Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

detection method

colorimetric

relevant disease(s)

neurological disorders; immunological diseases; hematological disorder

storage temp.

−20°C

General description

Purine nucleoside phosphorylase (PNP) (E.C. 2.4.2.1.) is an enzyme involved in purine metabolism and it catalyzes the cleavage of the glycosidic bond of ribo- or deoxyribonucleosides, in the presence of inorganic phosphate as a second substrate, to generate the purine base and ribose-1-phosphate or deoxyribose-1-phosphate. It is one of the enzymes of the nucleotide salvage pathways that allow the cell to produce nucleotide monophosphates when the de novo synthesis pathway has been interrupted or is non-existent (as is the case in the brain). PNP is a cytosolic enzyme.

Suitability

Suitable for the detection of Purine Nucleoside Phosphorylase activity in cell and tissue lysate or purified recombinant protein.

Principle

In this Purine Nucleoside Phosphorylase Activity Assay, hypoxanthine formed by the breakdown of inosine is further converted to uric acid using a developer. The uric acid is measured at a wavelength of 293 nm. Limit of quantification is 0.1μU recombinant purine nucleoside phosphorylase.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service