Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK283

Sigma-Aldrich

Myeloperoxidase (MPO) Inhibitor Screening Kit (Fluorometric)

sufficient for 200 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

detection method

fluorometric

relevant disease(s)

cardiovascular diseases; cancer

storage temp.

−20°C

General description

Myeloperoxidase (MPO: EC 1.11.2.2) is a peroxidase abundantly expressed in neutrophil granulocytes. It catalyzes the hydrogen peroxide dependent oxidation of halide ions to generate hypochlorite (HClO), the reaction by which MPO exhibits cytotoxic activity against tumor cells and microorganisms. MPO also oxidizes various substances such as phenol and anilines. MPO undergoes chlorination or peroxidation reaction depending upon the relative concentrations of chloride and the reducing substrates. Recent studies suggest that increased levels of MPO are associated with an increased risk for cardiovascular disease and myocardial infarction, thus development of novel and specific inhibitors of MPO is critical for therapeutic purposes.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for screening/characterizing/studying potential inhibitors of Myeloperoxidase

Principle

This MPO Inhibitor Screening Kit provides screening assays for both MPO chlorination and peroxidation activities. In the chlorination assay (specific for MPO), MPO converts hydrogen peroxide and sodium chloride to sodium hypochlorite, which reacts with the chlorination substrate to give an intensely fluorescent product, ex = 480 nm/em = 520. In the peroxidation assay, MPO oxidizes peroxidation substrate to generate fluorescence ex = 535 nm/em = 587. The fluorescence generated is directly proportional to any peroxidase activity present. In the presence of MPO inhibitor, reactions are impeded, thus decreasing the rate and/or extent of generation of MPO-dependent fluorescence.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service