Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK258

Sigma-Aldrich

Hepatic Steatosis Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

application(s)

pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

gastrointestinal diseases

storage temp.

−20°C

General description

Hepatic Steatosis, also known as fatty liver, is a disease characterized by an excessive accumulation of lipid droplets within the liver. This can be caused by multiple factors, including excessive alcohol intake, obesity, and other diseases with abnormal fat metabolism. Hepatic Steatosis is commonly used as a marker to determine the hepatotoxicity of drug candidates.

Suitability

Evaluate hepatic steatosis, quantify triglycerides, and visualize lipid droplet accumulation in hepatocytes. As well as screen drug candidates for steatotic risk.

Principle

The Hepatic Steatosis Kit detects triglyceride accumulation and lipid droplet formation in hepatocytes. The kit can be used to screen and evaluate steatosis risk of drug candidates and to study the mechanism of steatosis development. Chloroquine is included as a positive control for inducing steatosis in liver cells, such as HepG2.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service