Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK243

Sigma-Aldrich

Thrombin Inhibitor Screening Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

detection method

fluorometric

relevant disease(s)

hematological disorder

storage temp.

−20°C

General description

Thrombin enzyme (Factor IIa) is an important clotting factor that controls the transformation of soluble fibrinogen to insoluble active fibrin strands. Thrombin is a serine protease (EC 3.4.21.5) that catalyzes many coagulation-related reactions. Thrombin inhibitors are used as anticoagulants to prevent arterial and venous thrombosis. Some of these inhibitors are currently in clinical use while others are in clinical development.

Suitability

Suitable for screening potential inhibitors of thrombin and characterizing/studying thrombin inhibitors in plasma samples.

Principle

This Thrombin Inhibitor Screening Kit utilizes the ability of thrombin to cleave a synthetic AMC-based peptide substrate to release AMC, which can be detected by measuring its fluorescence (λex = 350 nm/λem = 450 nm). In the presence of thrombin specific inhibitors, the cleavage reaction is reduced or completely abolished. The loss in the fluorescence intensity can be correlated to the amount of inhibitor present in the assay solution. The kit provides a simple and rapid method to screen potential inhibitors of thrombin.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srinivas Chamakuri et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(36) (2021-08-25)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has killed more than 4 million humans globally, but there is no bona fide Food and Drug Administration-approved drug-like molecule to impede the COVID-19 pandemic. The sluggish pace of traditional therapeutic discovery is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service