Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK240

Sigma-Aldrich

Factor IXa Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

detection method

fluorometric

relevant disease(s)

hematological disorder

storage temp.

−20°C

General description

The coagulation Factor IX (or Christmas factor) (EC 3.4.21.22) is a vitamin K-dependent serine protease. Factor IX is produced as an inactive precursor and is activated via cleavage by either factor XIa (contact pathway) or factor VIIa (tissue factor pathway). In the presence of calcium ions and negatively charged membrane phospholipids, activated factor IX (FIXa) then binds to the activated Factor VIII (FVIIIa) and proteolytically activates factor X (FX) to factor Xa (FXa). Deficiency in factor IX causes hemophilia B, an X-linked bleeding disorder.[1]

Suitability

Suitable to determine the activity of purified Factor IX and to detect the activity of Factor IX in plasma and serum.

Principle

This Factor IXa Activity Assay kit is based on the ability of FIXa to generate FXa. The generated FXa proteolytically cleaves a synthetic substrate and releases a fluorophore, AMC, which can be easily quantified by fluorescence microplate reader. The assay is simple, rapid, and can detect activity as low as 10 pg of FIXa in a variety of samples.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenovirus-associated virus vector--mediated gene transfer in hemophilia B
Nathwani A C. et al.
The New England Journal of Medicine, 365(25), 2357-2365 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service