β-Secretase has been implicated to be an excellent target for anti-amyloid therapy for the treatment of Alzheimer′s disease.
Suitability
Suitable for the determination of ß-Secretase activity in biological and purified samples.
Principle
The β-Secretase activity Assay Kit provides a convenient fluorescence method for detecting β-secretase activity in biological and purified samples. The assay utilizes a secretase-specific peptide conjugated to two reporter molecules EDANS and DABCYL. In the uncleaved form, the fluorescent emissions from EDANS are quenched by the physical proximity of the DABCYL moiety. Cleavage of the peptide by secretase physically separates EDANS and DABCYL allowing for the release of a fluorescent signal. The level of secretase enzymatic activity in samples is proportional to the level of fluorescence intensity.
International journal of molecular sciences, 23(18) (2022-09-24)
Alzheimer's disease (AD), a common form of dementia, is caused in part by the aggregation and accumulation in the brain of amyloid β (Aβ), a product of the proteolytic cleavage of amyloid precursor protein (APP) in endosomes. Trafficking of APP
Questions
Reviews
★★★★★ No rating value