Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK236

Sigma-Aldrich

Factor VIIIa Activity Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

detection method

fluorometric

relevant disease(s)

hematological disorder

storage temp.

−20°C

General description

The coagulation Factor VIII, also known as anti-hemophilic factor (AHF) is a vital blood-clotting protein. Factor VIII circulates in the bloodstream as an inactive protein, bound to a large multimeric glycoprotein called von Willebrand factor (VWF). Upon an injury to the blood vessel, VWF dissociates from FVIII and releases the active form of FVIII (FVIIIa). In the presence of calcium ions and negatively charged membrane phospholipids, activated factor VIII (FVIIIa) then binds to the activated Factor IX (FIXa) and proteolytically activates factor X (FX) to factor Xa (FXa).
Acquired factor VIII deficiency results in skin, muscle and tissue hemorrhage.[1]

Suitability

Suitable for the determination of Factor VIII activity in serum and plasma samples

Principle

This Factor VIIIa Activity Assay kit is based on the ability of FVIIIa to generate FXa. The generated FXa proteolytically cleaves a synthetic substrate and releases a fluorophore, AMC, which can be easily quantified by a fluorescence microplate reader. The assay is simple, rapid, and can detect activity as low as 1 ng of FVIIIa in a variety of samples.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autoantibodies to coagulation factors: from pathophysiology to diagnosis and therapy
Cugno M, et al.
Autoimmunity Reviews, 13(1), 40-48 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service