Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK217

Sigma-Aldrich

Succinyl-CoA Synthetase Assay Kit

Sufficient for 100 Colorimetric tests

Synonym(s):

Succinate Thiokinase Assay Kit, succinyl-CoA ligase Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 reactions (Colorimetric)

detection method

colorimetric

storage temp.

−20°C

General description

Succinyl-CoA synthetase (SCS; succinyl-CoA ligase; succinate thiokinase; EC 6.2.1.5), also known as succinyl-CoA synthetase ligase/succinate thiokinase, is a mitochondrial enzyme. The enzyme catalyzes a substrate-level phosphorylation of GDP or ADP and thus, plays a vital role in Krebs cycle. In addition, it also catalyzes the reverse reaction to supply succinyl-CoA for heme synthesis and ketone body activation. Deficiency in SCS activity is associated with encephalomyopathy and depletion of mitochondrial DNA.Aberrations in the gene expression leads to the development of diabetes and neurodegenerative diseases.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of Succinly-CoA synthetase in a variety of biological samples such as cell and tissue culture supernatants and purified mitochondria.

Principle

The Succinyl-CoA Synthetase Activity Colorimetric Assay Kit is a simple and high throughput adaptable assay for measuring SCS activity in a variety of tissues, cells, and isolated mitochondria. SCS activity is determined by measuring a colorimetric product with absorbance at 450 nm (A450) proportional to the enzymatic activity present. One unit of succinyl-CoA synthetase is the amount of enzyme that generates 1.0 μmole of NADH per minute at pH 7.4 at 25 °C.

Kit Components Only

Product No.
Description

  • SCS Assay Buffer

  • SCS Substrate Mix

  • SCS Enzyme Mix

  • SCS Developer

  • NADH Standard

  • SCS Positive Control

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the ATP-and GTP-specific Succinyl-CoA Synthetases in Pigeon THE ENZYMES INCORPORATE THE SAME α-SUBUNIT.
Johnson, J. D,
The Journal of Biological Chemistry, 273(42), 27573-27579 (1998)
Deficiency of the ADP-Forming Succinyl-CoA Synthase Activity Is Associated with Encephalomyopathy and Mitochondrial DNA Depletion
Orly E, et al.
American Journal of Human Genetics, 76(6), 1081-1086 (2005)
Abby J Chiang et al.
mSystems, 4(4) (2019-06-13)
Bacillus pumilus SAFR-032 was originally isolated from the Jet Propulsion Lab Spacecraft Assembly Facility and thoroughly characterized for its enhanced resistance to UV irradiation and oxidative stress. This unusual resistance of SAFR-032 is of particular concern in the context of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service