Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK200

Sigma-Aldrich

Cathepsin B Inhibitor Screening Kit

Sufficient for 100 Fluorometric tests

Synonym(s):

CTSB Inhibitor Screening Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

detection method

fluorometric

storage temp.

−20°C

Gene Information

human ... CTSB(1508)
mouse ... CTSB(13030)
rat ... CTSB(64529)

General description

Cathepsin B (CTSB; EC 3.4.22.1) is a lysosomal cysteine proteinase involved in intracellular degradation and turnover of proteins. It is involved in the proteolytic processing of amyloid precursor protein (APP). Incomplete proteolytic processing of APP is a causative factor in Alzheimer′s disease. CTSB has also been implicated in the invasion and metastasis of certain cancers.

The Cathepsin B Inhibitor Screening Kit is a simple assay for screening potential inhibitors of CTSB. CTSB activity is measured by cleaving a synthetic 7-amino-4-trifluoromethylcoumarin (AFC)-based substrate to yield AFC, a fluorescent product (λex = 400/λem = 505 nm), proportional to the enzymatic activity present.

Suitability

Suitable for the screening of inhibitors of Cathepsin B.

Principle

The Cathepsin B Inhibitor Screening Kit is a simple assay for screening potential inhibitors of CTSB. CTSB activity is measured by cleaving a synthetic 7-amino-4-trifluoromethylcoumarin (AFC)-based substrate to yield AFC, a fluorescent product (λex = 400/λem = 505 nm), proportional to the enzymatic activity present.

Kit Components Only

Product No.
Description

  • CTSB Reaction Buffer

  • CTSB Reagent

  • Cathepsin B, human

  • CTSB Substrate, Ac-RR-AFC, 10 mM

  • CTSB Inhibitor, F-F-FMK, 1 mM

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Paoli et al.
Nature, 607(7917), 111-118 (2022-06-23)
Natural microbial communities are phylogenetically and metabolically diverse. In addition to underexplored organismal groups1, this diversity encompasses a rich discovery potential for ecologically and biotechnologically relevant enzymes and biochemical compounds2,3. However, studying this diversity to identify genomic pathways for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service