Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK149

Sigma-Aldrich

NIR Mitochondrion Membrane Potential Kit

sufficient for 500 fluorometric tests (near infrared fluorescence, microplate readers)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 500 fluorometric tests (near infrared fluorescence, microplate readers)

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Mitochondria generate a potential across their membranes due to the activities of enzymes of the electron transport chain. During apoptosis, collapse of the mitochondrial membrane potential (MMP) coincides with the opening of the mitochondrial permeability transition pores, leading to the release of cytochrome c into the cytosol, which in turn triggers other downstream events in the apoptotic cascade.

Suitability

This kit is suitable for the detection of Mitochondrial Membrane Potential in mammalian cells and for screening apoptosis inhibitors and activators using microplate readers.

Principle

NIR Mitochondrion Membrane Potential Kit is optimized for the detection of the loss of the MMP in cells. The cationic hydrophobic mitochondrial potential dye accumulates in normal mitochondria, most likely due to the mitochondrial potential, resulting in an increase in fluorescence (λex = 640/λem = 680 nm). In apoptotic cells, MMP collapse results in decreased fluorescence. This kit can be used for monitoring apoptosis and for screening apoptosis inhibitors and activators.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service