Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK146

Sigma-Aldrich

Mitochondrion Membrane Potential Kit

sufficient for 100 fluorometric tests (flow cytometry, orange fluorescence)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 100 fluorometric tests (flow cytometry, orange fluorescence)

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Mitochondria generate a potential across their membranes due to the activities of enzymes of the electron transport chain. During apoptosis, collapse of the mitochondrial membrane potential (MMP) coincides with the opening of the mitochondrial permeability transition pores, leading to the release of cytochrome c into the cytosol, which in turn triggers other downstream events in the apoptotic cascade.[1][2]

Suitability

This kit is suitable for the flow cytometric detection of Mitochondrial Membrane Potential in mammalian cells and for screening apoptosis inhibitors and activators.

Principle

This kit is optimized for the analysis of the MMP by flow cytometry on instruments with a 488 nm laser. The cationic hydrophobic mitochondrial potential dye accumulates in normal mitochondria, most likely due to the mitochondrial potential, resulting in an increase in fluorescence (λem = 535 nm). In apoptotic cells, MMP collapse results in decreased fluorescence. This kit can be used for monitoring apoptosis and for screening apoptosis inhibitors and activators.
This kit is optimized for the analysis of the MMP by flow cytometry on instruments with a 488 nm laser. The cationic hydrophobic mitochondrial potential dye accumulates in normal mitochondria, most likely due to the mitochondrial potential, resulting in an increase in fluorescence (λem = 535 nm). In apoptotic cells, MMP collapse results in decreased fluorescence. This kit can be used for monitoring apoptosis and for screening apoptosis inhibitors and activators.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitochondrial membrane permeabilization and cell death during myocardial infarction: roles of calcium and reactive oxygen species.
Webster K A
Future Cardiology, 8(6), 863-884 (2012)
Role of mitochondrial permeability transition pores in mitochondrial autophagy.
Rodriguez-Enriquez S, et al.
The International Journal of Biochemistry & Cell Biology, 36(12), 2463-2472 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service