Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAK103

Sigma-Aldrich

Phosphoglucose Isomerase Colorimetric Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

storage temp.

−20°C

General description

Phosphoglucose Isomerase (PGI) is an enzyme crucial for the interconversion of D-glucose 6-phosphate and D-fructose 6-phosphate in the second step of glycolysis and is involved in glucogenesis.[1] PGI is also referred to as Autocrine Motility Factor (AMF) and is secreted by cancer cells in order to stimulate metastasis.[2] Deficiencies in PGI activity can lead to hemolytic anemia.[1]

Suitability

Suitable for the measurement of phosphoglucose isomerase activity, analysis of glucose metabolism and cell signaling and screening anti-diabetic drugs in animal tissues (liver, brain, heart, kidney etc.), cell culture (adherent or suspension cells), bacteria, yeast, fish etc.

Principle

The Phosphoglucose Isomerase Colorimetric Assay kit provides a simple and direct procedure for measuring PGI activity in a variety of samples. PGI activity is determined by a coupled enzyme assay in which fructose-6-phosphate is converted by PGI to glucose-6-phosphate. Glucose-6-phosphate is subsequently oxidized to form a product, which reacts with a probe generating a colorimetric (450 nm) product proportional to the PGI activity present. One unit of PGI is the amount of enzyme that will generate 1.0 μmole of NADH per minute at pH 8.0 at room temperature.

pictograms

Health hazardCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of rabbit phosphoglucose isomerase complexed with its substrate D-fructose 6-phosphate.
Lee J H, et al.
Biochemistry, 40(26), 7799-7805 (2001)
Seok Hyun Park et al.
Nature communications, 5, 4618-4618 (2014-08-06)
L-arginine is an important amino acid for diverse industrial and health product applications. Here we report the development of metabolically engineered Corynebacterium glutamicum ATCC 21831 for the production of L-arginine. Random mutagenesis is first performed to increase the tolerance of
Metabolic engineering of Corynebacterium glutamicum for L-arginine production.
Park S H, et al.
Nature Communications, 5, 4618-4618 (2014)
Regulation of cell proliferation by autocrine motility factor/phosphoglucose isomerase signaling.
Tsutsumi S, et al.
The Journal of Biological Chemistry, 278(34), 32165-32172 (2003)
Gabriela Krejčová et al.
eLife, 8 (2019-10-15)
Macrophage-mediated phagocytosis and cytokine production represent the front lines of resistance to bacterial invaders. A key feature of this pro-inflammatory response in mammals is the complex remodeling of cellular metabolism towards aerobic glycolysis. Although the function of bactericidal macrophages is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service