Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK101

Sigma-Aldrich

PAH (p-Aminohippuric Acid) Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

genitourinary disorders

storage temp.

−20°C

General description

p-Aminohippuric acid (PAH), a derivative of hippuric acid, is used in the measurement of renal plasma flow (RPF). Renal plasma flow measures the volume of plasma, which enters the kidney in a given period of time. To determine effective renal plasma flow, PAH is measured in a plasma sample (PPAH) and a urine sample (UPAH). The volume of urine flow (V) is also required.[1][2]

Suitability

Suitable for measuring PAH (p-Aminohippuric Acid) in plasma and urine samples

Principle

The PAH (p-Aminohippuric Acid) Assay kit can be used for measuring PAH in a plasma and urine samples. PAH reacts with 4-(dimethylamino)cinnamaldehyde (DACA) to generate a compound, which can be detected spectrophotometrically at 550 nm.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumor necrosis factor induces glomerular damage in the rabbit.
Bertani T, et al.
The American Journal of Pathology, 134(2), 419-419 (1989)
Evaluation of change in blood flow by contrast?enhanced power Doppler imaging during norepinephrine?induced renal vasoconstriction.
Arger P H, et al.
Journal of Ultrasound in Medicine : Official Journal of the American Institute of Ultrasound in Medicine, 18(12), 843-851 (1999)
Cesar A Romero et al.
American journal of physiology. Renal physiology, 316(1), F195-F203 (2018-11-08)
The antifibrotic peptide N-acetyl-seryl-aspartyl-lysyl-proline (Ac-SDKP) is released from thymosin-β4 (Tβ4) by the meprin-α and prolyl oligopeptidase (POP) enzymes and is hydrolyzed by angiotensin-converting enzyme (ACE). Ac-SDKP is present in urine; however, it is not clear whether de novo tubular release

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service