Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK073

Sigma-Aldrich

Sarcosine Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

relevant disease(s)

neurological disorders

storage temp.

−20°C

General description

Sarcosine, also known as N-methylglycine, is an amino acid that participates in one-carbon metabolism, serving as a one-carbon donor in mitochondrial reactions. Elevated levels of sarcosine may indicate deficiencies in Vitamin B12 and/or folate. Sarcosine is also an intermediate in glycine and choline metabolism. In the brain, sarcosine acts as an inhibitor of the type I glycine transporter, resulting in increased glycine levels and NMDA receptor activation suggesting it may be a therapeutic agent for the treatment of some mental illnesses such as schizophrenia, depression, or obsessive compulsive disorder.

Suitability

This kit is suitable for sarcosine detection in cell and tissue culture supernatants, plasma, serum, and other biological samples. Urine samples are not suitable for use with this assay because of sample interferences.

Principle

In this assay, sarcosine concentration is determined by a coupled enzyme reaction, which results in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the sarcosine present.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joan Artigas et al.
Environmental pollution (Barking, Essex : 1987), 259, 113951-113951 (2020-01-16)
Environmental dissolved organic matter (DOM) has been proved to increase microbial population sizes and stimulate the degradation of some pesticide molecules. Among these molecules, the present study investigated the biodegradation of the herbicide glyphosate depending on photoautotrophs DOM supply in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service