Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

MABD690

Sigma-Aldrich

SOX 9 antibody

enhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-SOX 9 antibody, Transcription factor SOX-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B10, monoclonal

mol wt

calculated mol wt 56.14 kDa
observed mol wt ~65 kDa (Uncharacterized bands may be observed in some lysate(s).)

species reactivity

human, mouse

species reactivity (predicted by homology)

rat

packaging

pkg of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

western blot: 1:1000

isotype

IgG

Storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SOX9(6662)

General description

Transcription factor SOX-9 (UniProt P48436; also known SOX-9) is encoded by the SOX9 gene (Gene ID: 6662) in human. SOX family of genes are related to the mammalian sex determining gene SRY. These genes contain sequences encoding for the HMG-box domains that mediate sequence-specific DNA binding. SOX genes encode transcriptional regulators involved in cell fate decision during development and the control of diverse developmental processes. The highly complex group of SOX genes cluster at about 40 genomic loci and at least 30 SOX genes have been identified. SOX-9 contains one HMG box DNA-binding domain and acts as a key regulator of developmental processes, including male sex determination, normal skeletal development, chondrogenesis, neurogenesis, and neural crest development. SOX-9 is acts downstream of sonic hedgehog signaling and is shown to be essential for the maintenance of multipotent neural stem cells in embryonic and adult central nervous system (CNS). It is not detected in neuroblasts of the adult subependymal zone (SEZ); however, it is expressed by transit amplifying cells and ependymal cells in the same region. SOX-9 is also shown to be essential for the ′switch′ from neurogenesis to gliogenesis in caudal regions of the developing CNS. Up-regulation of SOX-9 is reported in cancers of various origins, including breast, colon, prostate, and lung, where its expression correlates with malignancy and patient survival. This recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Scott, CE et al. (2010). Nat. Neurosci. 13(10); 1181-89).

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal region of human SOX-9.

Application

Anti-SOX-9 Antibody, clone 2B10, Cat. No. MABD690, is a rabbit monoclonal antibody that detects Transcription factor SOX-9 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Physical form

Lyophilized, resin-like pellet. Purified recombinant rabbit monoclonal antibody IgG lyophilized from PBS with 5% Trehalose. (Reconstitute with 25 μL of distilled water). No azide or preservatives added. Larger pack sizes will be provided as multiples of 25 μL.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service