Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9693

Sigma-Aldrich

Anti-Muscarinic Acetylcholine Receptor M2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 2 μg/mL using brain tissue, neurons

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRM2(1129)
mouse ... Chrm2(243764)
rat ... Chrm2(81645)

General description

Muscarinic acetylcholine receptor M2 is expressed in the heart and many other tissues. The gene encoding it is localized on human chromosome 7.

Immunogen

synthetic peptide corresponding to the third cytoplasmic loop of human muscarinic acetylcholine receptor M2, conjugated to KLH. The immunizing peptide has 100% homology with the mouse and rat gene.

Biochem/physiol Actions

Muscarinic acetylcholine receptor M2 plays an important role in regulating functions of the heart and processes like pain perception and cognition. It is regarded as a good model in G protein-coupled receptors (GPCR) pharmacological studies. This receptor has a function in airway remodeling and it enhances the proliferation of airway smooth muscle cells (ASM) which is induced by transforming growth factor-β (TGF-β).

Physical form

Solution in phosphate buffered saline, containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew C Kruse et al.
Nature, 504(7478), 101-106 (2013-11-22)
Despite recent advances in crystallography and the availability of G-protein-coupled receptor (GPCR) structures, little is known about the mechanism of their activation process, as only the β2 adrenergic receptor (β2AR) and rhodopsin have been crystallized in fully active conformations. Here
Yinglong Miao et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(27), 10982-10987 (2013-06-20)
G-protein-coupled receptors (GPCRs) mediate cellular responses to various hormones and neurotransmitters and are important targets for treating a wide spectrum of diseases. Although significant advances have been made in structural studies of GPCRs, details of their activation mechanism remain unclear.
Shirley Y Hill et al.
Psychiatry research, 209(3), 504-511 (2013-06-12)
P300 amplitude in childhood predicts substance use disorders by young adulthood. Trajectories of visual P300 amplitude show an association between low amplitude P300 and familial risk for alcohol dependence (AD). Variation in the cholinergic muscarinic receptor gene (CHRM2) has previously
Tjitske A Oenema et al.
American journal of respiratory cell and molecular biology, 49(1), 18-27 (2013-03-02)
Transforming growth factor-β₁ (TGF-β₁) is a central mediator in tissue remodeling processes, including fibrosis and airway smooth muscle (ASM) hyperplasia, as observed in asthma. The mechanisms underlying this response, however, remain unclear because TGF-β₁ exerts only weak mitogenic effects on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service