Muscarinic acetylcholine receptor M3 possesses a vestibule which is a part of a hydrophilic channel having a orthosteric binding site.
Specificity
Muscarinic acetylcholine receptor m3.
Immunogen
Synthetic peptide from the third extracellular loop of rat m3 (Accession P08483).
Biochem/physiol Actions
Muscarinic acetylcholine receptor M3 functions as a protectant in myocardial ischemia. It also has roles in smooth muscle contraction and glandular secretion. The receptor has a specific affinity for the Gq/11 family of G proteins.
Target description
Muscarinic Acetylcholine Receptor M3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue.
Physical form
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Clinical and experimental pharmacology & physiology, 47(2), 286-293 (2019-10-21)
Here, we investigate the effects of exercise training on glucose- and cholinergic-induced insulin secretion in pancreatic islets from obese and lean rats. Male Wistar rats were treated with monosodium glutamate (MSG) for the first 5 days of life, while control (CON)
Acetylcholine, the first neurotransmitter to be identified, exerts many of its physiological actions via activation of a family of G-protein-coupled receptors (GPCRs) known as muscarinic acetylcholine receptors (mAChRs). Although the five mAChR subtypes (M1-M5) share a high degree of sequence
The M(3) subtype of muscarinic acetylcholine receptors (M(3)-mAChR) plays a protective role in myocardial ischemia and microRNAs (miRNAs) participate in many cardiac pathophysiological processes, including ischemia-induced cardiac injury. However, the role of miRNAs in M(3)-mAChR mediated cardioprotection remains unexplored. The
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.