Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9447

Sigma-Aldrich

MRCKα (1-473), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDC42BPA, DKFZp686L1 738, DKFZp686P1 738, FLJ23347, KIAA0451, PK428

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

86-116 nmol/min·mg

mol wt

~73 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CDC42BPA(8476)

Biochem/physiol Actions

Myotonic Dystrophy Kinase-Related Cdc42-Binding Kinase a (MRCKa) is a Cdc42/Rac/Rho interactive/binding serine/threonine kinase with multiple functional domains. MRCK are effectors of RhoA and Cdc42, respectively, for actin reorganization. MRCKa is a critical regulator of signal transduction pathways in eukaryotic cells that are known principally for their role in regulating the cytoskeleton, and they do so by recruiting a variety of downstream effector proteins.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan Tan et al.
Gene, 304, 107-115 (2003-02-06)
Myotonic dystrophy kinase-related Cdc42-binding kinase alpha (MRCKalpha) is a Cdc42/Rac interactive binding-containing serine/threonine kinase with multiple functional domains. Its roles in the regulation of peripheral actin reorganization in HeLa cells and NGF-induced neurite outgrowth in PC12 cells have been documented.
I Tan et al.
The Journal of biological chemistry, 276(24), 21209-21216 (2001-06-16)
The myotonic dystrophy kinase-related kinases RhoA binding kinase and myotonic dystrophy kinase-related Cdc42 binding kinase (MRCK) are effectors of RhoA and Cdc42, respectively, for actin reorganization. Using substrate screening in various tissues, we uncovered two major substrates, p130 and p85

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service