Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9322

Sigma-Aldrich

MNK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

GPRK7, MKNK2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$403.00

$403.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$403.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$403.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

81-109 nmol/min·mg

mol wt

~74 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MKNK2(2872)

Biochem/physiol Actions

MNK2 (MAP kinase-interacting kinase 2) contains a conserved C-terminal ERK-interacting domain, a catalytic domain with homology to the calcium/calmodulin-dependent family of kinases, and putative MAP kinase phosphorylation sites located within the T loop of the kinase domain. MNK2 binds tightly to the growth factor-regulated MAP kinases, ERK1 and ERK2. ERK and p38 phosphorylate MNK2, which stimulates its in vitro kinase activity toward a substrate, eukaryotic initiation factor-4E (eIF-4E).A yeast two-hybrid screen showed the MNK2 protein interacted with the ligand-binding domain of estrogen receptor beta (ERbeta).

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Slentz-Kesler et al.
Genomics, 69(1), 63-71 (2000-10-03)
We have identified and characterized the human Mnk2 gene (HGMW-approved gene symbol MKNK2) through a yeast two-hybrid screen in which the Mnk2 protein interacted with the ligand-binding domain of estrogen receptor beta (ERbeta). Human Mnk2 is homologous to murine Mnk2
A J Waskiewicz et al.
The EMBO journal, 16(8), 1909-1920 (1997-04-15)
Mitogen-activated protein (MAP) kinases bind tightly to many of their physiologically relevant substrates. We have identified a new subfamily of murine serine/threonine kinases, whose members, MAP kinase-interacting kinase 1 (Mnk1) and Mnk2, bind tightly to the growth factor-regulated MAP kinases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service