Matrix metalloproteinase-21 (MMP-21) is a secreted protein expressed in fetal tissues, adult tissues and cancer cell lines. It is a 57kDa protein. The gene encoding it is localized on human chromosome 10.
Specificity
By immunoblotting against the reduced protein, the antibody identifies bands at 70 kDa and 66 kDa.
Immunogen
synthetic peptide corresponding to the hinge region of human matrix metalloproteinase-21.
Biochem/physiol Actions
Matrix metalloproteinase-21 (MMP-21) is involved in increasing the invasion capacity and metastasis property of tumors.
Physical form
Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
During intracellular infections, autophagy significantly contributes to the elimination of pathogens, regulation of pro-inflammatory signaling, secretion of immune mediators and in coordinating the adaptive immune system. Intracellular pathogens such as S. Typhimurium have evolved mechanisms to circumvent autophagy. However, the
We have characterized a novel human matrix metalloproteinase (MMP-21) from human placenta DNA complementary to RNA (cDNA). The 569 amino acid translation of the cDNA includes all the typical features of an MMP family member, namely a signal sequence, a
Matrix metalloproteinase (MMP)-21 and MMP-26 (matrilysin-2) are two recently cloned epithelial metalloproteases. Here we examined their expression in various benign skin disorders, in which macrophages and fibroblasts have been implicated as well as in cultures of these cells. Expression of
Medical oncology (Northwood, London, England), 30(1), 323-323 (2013-01-01)
Matrix metalloproteinase-21 (MMP-21) has been shown to enhance tumor invasion and metastasis ability in some solid tumors. In the present study, we investigated the expression of MMP-21 as well as its association with overall survival of gastric cancer patients. MMP-21
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.