Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8572

Sigma-Aldrich

MARK3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CTAK1, KP78, PAR1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

614-830 nmol/min·mg

mol wt

~110 kDa

UniProt accession no.

shipped in

dry ice

Storage temp.

−70°C

Gene Information

human ... MARK3(4140)

Biochem/physiol Actions

Microtubule affinity-regulating kinase 3 (MARK3) is a member of the PAR-1/MARK kinase family which play critical roles in polarity and cell cycle control and are regulated by 14-3-3 scaffolding proteins, as well as the LKB1 tumour suppressor kinase and atypical protein kinase C (PKC). MARK3 is a dual-specificity protein kinase that controls entry into mitosis by dephosphorylating CDC2. MARK3 seems to be a positive regulator of the beta-catenin pathway and an inhibitor of the JNK pathway. MARK3, a regulator of polarity, is also a modulator of Wnt-beta-catenin signalling, indicating a link between two important developmental pathways.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kato et al.
Neoplasia (New York, N.Y.), 3(1), 4-9 (2001-04-28)
Activation of the Wnt-signaling pathway is known to play a crucial role in carcinogenesis of various human organs including the colon, liver, prostate, and endometrium. To investigate the mechanisms underlying hepatocellular carcinogenesis, we attempted to identify genes regulated by beta-catenin/Tcf
Olga Göransson et al.
Journal of cell science, 119(Pt 19), 4059-4070 (2006-09-14)
Members of the PAR-1/MARK kinase family play critical roles in polarity and cell cycle control and are regulated by 14-3-3 scaffolding proteins, as well as the LKB1 tumour suppressor kinase and atypical protein kinase C (PKC). In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service